Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01794397_g1
See other CAP GE Assays ›
Gene Symbol:
CAP
Gene Name:
CAP-PA
Gene Aliases:
CAP/Vinexin, CG18408, CG18409, CG3451, DCAP, Dmel\CG18408, Dmel_CG18408, Rexin, cap, rexin
Chromosome Location:
Chr.2R: 10269016 - 10307024 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0033504
Assay ID
Dm01794397_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CAP
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001144164.2
NP_001137636.1
10 - 11
1282
84
NM_001144165.2
NP_001137637.1
4 - 5
6395
84
NM_001144166.3
NP_001137638.2
6 - 7
6635
84
NM_001144168.2
NP_001137640.1
8 - 9
805
84
NM_001169639.2
NP_001163110.1
4 - 5
451
84
NM_001259309.2
NP_001246238.1
5 - 6
650
84
NM_001259310.2
NP_001246239.2
7 - 8
6815
84
NM_001259311.2
NP_001246240.1
9 - 10
853
84
NM_001259312.2
NP_001246241.1
8 - 9
6674
84
NM_001273930.1
NP_001260859.1
4 - 5
3509
84
NM_001273931.1
NP_001260860.1
6 - 7
6527
84
NM_001382041.1
NP_001369073.1
11 - 12
1560
84
NM_001382042.1
NP_001369074.1
6 - 7
6965
84
NM_001382043.1
NP_001369075.1
8 - 9
7004
84
NM_001382044.1
NP_001369076.1
4 - 5
3839
84
NM_001382045.1
NP_001369077.1
6 - 7
6857
84
NM_136726.3
NP_610570.1
7 - 8
644
84
NM_136727.4
NP_610571.2
5 - 6
6428
84
NM_136728.4
NP_610572.1
5 - 6
559
84
NM_165757.3
NP_724910.1
11 - 12
1303
84
NM_165758.2
NP_724911.1
10 - 11
1491
84
NM_165759.3
NP_724912.1
5 - 6
1502
84
NM_165760.4
NP_724913.2
4 - 5
451
84
NM_206073.3
NP_995795.1
6 - 7
683
84
GenBank mRNA
AB053479.1
-
4 - 5
435
84
AY061565.1
-
5 - 6
526
84
BT003217.1
-
11 - 12
1304
84
BT003435.1
-
10 - 11
1263
84
BT120189.1
-
5 - 6
643
84
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Drosophila melanogaster
Gene Ontology Categories:
Function(s)
sensory perception of sound
muscle attachment
sensory perception of mechanical stimulus
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0