Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp49a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259327.2 | NP_001246256.1 | 5 - 6 | 1094 | 67 | |
NM_136744.4 | NP_610588.2 | 5 - 6 | 854 | 67 | ||
NM_165774.3 | NP_724937.1 | 1 - 2 | 159 | 67 | ||
NM_206081.2 | NP_995803.1 | 5 - 6 | 800 | 67 | ||
GenBank mRNA | BT056244.1 | - | 5 - 6 | 846 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |