Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp49a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259327.2 | NP_001246256.1 | 8 - 9 | 1484 | 63 | |
NM_136744.4 | NP_610588.2 | 8 - 9 | 1244 | 63 | ||
NM_165774.3 | NP_724937.1 | 4 - 5 | 549 | 63 | ||
NM_206081.2 | NP_995803.1 | 8 - 9 | 1190 | 63 | ||
GenBank mRNA | BT056244.1 | - | 8 - 9 | 1236 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |