Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: luna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273950.1 | NP_001260879.1 | 2 - 3 | 1489 | 57 | |
NM_001299374.1 | NP_001286303.1 | 2 - 3 | 918 | 57 | ||
NM_206089.2 | NP_995811.1 | 2 - 3 | 1489 | 57 | ||
GenBank mRNA | AF461497.1 | - | 1 - 2 | 98 | 57 | |
AY122143.1 | - | 1 - 2 | 468 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |