Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: luna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273950.1 | NP_001260879.1 | 3 - 4 | 2782 | 52 | |
NM_001299374.1 | NP_001286303.1 | 3 - 4 | 2211 | 52 | ||
NM_206089.2 | NP_995811.1 | 3 - 4 | 2782 | 52 | ||
GenBank mRNA | AF461497.1 | - | 2 - 3 | 1391 | 52 | |
AY122143.1 | - | 2 - 3 | 1761 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |