Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nach | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347816.1 | NP_001334723.1 | 1 - 2 | 358 | 94 | |
NM_001347817.1 | NP_001334724.1 | 1 - 2 | 358 | 94 | ||
NM_137301.1 | NP_611145.1 | 1 - 2 | 358 | 94 | ||
GenBank mRNA | AY226538.1 | - | 1 - 2 | 358 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |