Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nach | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347816.1 | NP_001334723.1 | 4 - 5 | 1384 | 91 | |
NM_001347817.1 | NP_001334724.1 | 3 - 4 | 1438 | 91 | ||
NM_137301.1 | NP_611145.1 | 5 - 6 | 1327 | 91 | ||
GenBank mRNA | AY226538.1 | - | 4 - 5 | 1384 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |