Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mute | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103882.3 | NP_001097352.1 | 2 - 3 | 945 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |