Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144353.4 | NP_652610.1 | 2 - 3 | 479 | 104 | |
GenBank mRNA | AF222864.1 | - | 2 - 3 | 455 | 104 | |
BT023840.1 | - | 2 - 3 | 455 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |