Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014540.3 | NP_001014540.2 | 3 - 4 | 1844 | 102 | |
NM_001014542.3 | NP_001014542.1 | 3 - 4 | 1844 | 102 | ||
NM_001169737.2 | NP_001163208.1 | 3 - 4 | 1941 | 102 | ||
NM_001169738.2 | NP_001163209.1 | 3 - 4 | 1941 | 102 | ||
NM_001169739.2 | NP_001163210.1 | 3 - 4 | 1844 | 102 | ||
NM_001169740.2 | NP_001163211.1 | 3 - 4 | 1985 | 102 | ||
NM_206171.4 | NP_995893.1 | 3 - 4 | 1743 | 102 | ||
NM_206172.3 | NP_995894.2 | 3 - 4 | 1844 | 102 | ||
NM_206177.3 | NP_995899.2 | 3 - 4 | 1844 | 102 | ||
NM_206178.3 | NP_995900.1 | 3 - 4 | 1844 | 102 | ||
GenBank mRNA | AF258462.1 | - | 3 - 4 | 1819 | 102 | |
AF387635.1 | - | 3 - 4 | 1819 | 102 | ||
AF387636.1 | - | 2 - 3 | 1761 | 102 | ||
BT009940.1 | - | 3 - 4 | 1893 | 102 | ||
BT011020.1 | - | 3 - 4 | 1839 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |