Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259599.2 | NP_001246528.1 | 5 - 6 | 1433 | 83 | |
NM_138163.2 | NP_612007.2 | 5 - 6 | 1381 | 83 | ||
NM_206222.3 | NP_995944.1 | 5 - 6 | 1597 | 83 | ||
GenBank mRNA | AY321364.1 | - | 5 - 6 | 1380 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |