Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zormin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043112.2 | NP_001036577.2 | 13 - 14 | 7297 | 82 | |
NM_001043113.2 | NP_001036578.2 | 14 - 15 | 7369 | 82 | ||
NM_206240.2 | NP_995962.2 | 13 - 14 | 7297 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |