Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zormin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274386.1 | NP_001261315.1 | 17 - 18 | 10527 | 71 | |
NM_206240.2 | NP_995962.2 | 14 - 15 | 8724 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |