Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zormin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043112.2 | NP_001036577.2 | 6 - 7 | 2581 | 78 | |
NM_001043113.2 | NP_001036578.2 | 6 - 7 | 2581 | 78 | ||
NM_001043114.2 | NP_001036579.2 | 6 - 7 | 2581 | 78 | ||
NM_001274386.1 | NP_001261315.1 | 7 - 8 | 2629 | 78 | ||
NM_001274388.1 | NP_001261317.1 | 6 - 7 | 2581 | 78 | ||
NM_206240.2 | NP_995962.2 | 6 - 7 | 2581 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |