Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zormin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043112.2 | NP_001036577.2 | 8 - 9 | 3536 | 101 | |
NM_001043113.2 | NP_001036578.2 | 9 - 10 | 3608 | 101 | ||
NM_001043114.2 | NP_001036579.2 | 8 - 9 | 3536 | 101 | ||
NM_001274386.1 | NP_001261315.1 | 10 - 11 | 3656 | 101 | ||
NM_001274388.1 | NP_001261317.1 | 9 - 10 | 3608 | 101 | ||
NM_206240.2 | NP_995962.2 | 8 - 9 | 3536 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |