Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zormin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043112.2 | NP_001036577.2 | 10 - 11 | 5114 | 76 | |
NM_001043113.2 | NP_001036578.2 | 11 - 12 | 5186 | 76 | ||
NM_001043114.2 | NP_001036579.2 | 10 - 11 | 5114 | 76 | ||
NM_001274386.1 | NP_001261315.1 | 12 - 13 | 5234 | 76 | ||
NM_001274388.1 | NP_001261317.1 | 11 - 12 | 5186 | 76 | ||
NM_206240.2 | NP_995962.2 | 10 - 11 | 5114 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |