Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104022.3 | NP_001097492.2 | 12 - 13 | 12221 | 64 | |
NM_001202112.2 | NP_001189041.1 | 13 - 14 | 12283 | 64 | ||
NM_001274438.1 | NP_001261367.1 | 12 - 13 | 12221 | 64 | ||
NM_001274439.1 | NP_001261368.1 | 13 - 14 | 12249 | 64 | ||
NM_079176.3 | NP_523900.1 | 12 - 13 | 12718 | 64 | ||
NM_206265.3 | NP_995987.1 | 12 - 13 | 12718 | 64 | ||
NM_206266.3 | NP_995988.1 | 12 - 13 | 12718 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |