Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104046.4 | NP_001097516.3 | 6 - 7 | 4702 | 50 | |
NM_001104047.5 | NP_001097517.1 | 17 - 18 | 4462 | 50 | ||
NM_001104048.2 | NP_001097518.2 | 19 - 20 | 4122 | 50 | ||
NM_001104049.4 | NP_001097519.2 | 18 - 19 | 3917 | 50 | ||
NM_001274517.3 | NP_001261446.1 | 18 - 19 | 4519 | 50 | ||
NM_001274518.2 | NP_001261447.1 | 5 - 6 | 1390 | 50 | ||
NM_001274519.3 | NP_001261448.1 | 18 - 19 | 4492 | 50 | ||
NM_001300018.2 | NP_001286947.1 | 17 - 18 | 5474 | 50 | ||
NM_001347763.1 | NP_001334674.1 | 19 - 20 | 4438 | 50 | ||
NM_001347764.1 | NP_001334675.1 | 19 - 20 | 4474 | 50 | ||
NM_079908.3 | NP_524647.2 | 6 - 7 | 4702 | 50 | ||
NM_168126.4 | NP_729084.1 | 17 - 18 | 3472 | 50 | ||
NM_206278.2 | NP_996000.1 | 6 - 7 | 4702 | 50 | ||
GenBank mRNA | AY118305.1 | - | 6 - 7 | 4680 | 50 |
Gene Symbol: CG46320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347801.1 | NP_001334709.1 | 17 - 18 | 4462 | 50 | |
NM_001347802.1 | NP_001334710.1 | 19 - 20 | 4438 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |