Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259741.1 | NP_001246670.1 | 9 - 10 | 6249 | 88 | |
NM_001259742.1 | NP_001246671.1 | 9 - 10 | 6546 | 88 | ||
NM_001259743.1 | NP_001246672.1 | 9 - 10 | 6246 | 88 | ||
NM_001274651.1 | NP_001261580.1 | 9 - 10 | 6531 | 88 | ||
NM_001274652.1 | NP_001261581.1 | 10 - 11 | 6603 | 88 | ||
NM_079249.3 | NP_523973.2 | 10 - 11 | 6603 | 88 | ||
NM_145106.1 | NP_659574.1 | 9 - 10 | 6050 | 88 | ||
NM_176300.1 | NP_788478.1 | 10 - 11 | 6107 | 88 | ||
NM_206303.2 | NP_996025.1 | 10 - 11 | 6603 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |