Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259741.1 | NP_001246670.1 | 2 - 3 | 666 | 91 | |
NM_001259742.1 | NP_001246671.1 | 3 - 4 | 1020 | 91 | ||
NM_001259743.1 | NP_001246672.1 | 2 - 3 | 666 | 91 | ||
NM_001274652.1 | NP_001261581.1 | 3 - 4 | 1020 | 91 | ||
NM_079249.3 | NP_523973.2 | 3 - 4 | 1020 | 91 | ||
NM_145106.1 | NP_659574.1 | 3 - 4 | 524 | 91 | ||
NM_176300.1 | NP_788478.1 | 3 - 4 | 524 | 91 | ||
NM_206303.2 | NP_996025.1 | 3 - 4 | 1020 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |