Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tip60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272304.1 | NP_001259233.1 | 1 - 2 | 652 | 67 | |
NM_001272305.1 | NP_001259234.1 | 1 - 2 | 159 | 67 | ||
NM_131923.4 | NP_572151.1 | 1 - 2 | 697 | 67 | ||
GenBank mRNA | AY051795.1 | - | 1 - 2 | 652 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |