Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sas10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131987.4 | NP_572215.1 | 1 | 1457 | 141 | |
GenBank mRNA | AY047575.1 | - | 1 | 1427 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |