Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297991.1 | NP_001284920.1 | 10 - 11 | 3225 | 76 | |
NM_001297992.1 | NP_001284921.1 | 9 - 10 | 3176 | 76 | ||
NM_132057.3 | NP_572285.2 | 9 - 10 | 3363 | 76 | ||
GenBank mRNA | BT044143.1 | - | 9 - 10 | 3362 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |