Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca-alpha1T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103419.2 | NP_001096889.1 | 6 - 7 | 1646 | 53 | |
NM_001258615.1 | NP_001245544.1 | 6 - 7 | 1646 | 53 | ||
NM_001258616.1 | NP_001245545.1 | 6 - 7 | 1646 | 53 | ||
NM_001272338.1 | NP_001259267.1 | 6 - 7 | 1646 | 53 | ||
NM_132068.3 | NP_572296.2 | 6 - 7 | 1646 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |