Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201903.1 | NP_001188832.1 | 4 - 5 | 907 | 80 | |
NM_001201904.1 | NP_001188833.1 | 4 - 5 | 1165 | 80 | ||
NM_001299090.1 | NP_001286019.1 | 4 - 5 | 1165 | 80 | ||
NM_136009.2 | NP_609853.1 | 4 - 5 | 1013 | 80 | ||
GenBank mRNA | BT060450.1 | - | 4 - 5 | 907 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |