Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CadN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042903.4 | NP_001036368.2 | 4 - 5 | 792 | 110 | |
NM_136011.4 | NP_609855.3 | 5 - 6 | 1486 | 110 | ||
GenBank mRNA | BT132666.1 | - | 4 - 5 | 759 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |