Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042904.4 | NP_001036369.3 | 20 - 21 | 8111 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |