Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136033.3 | NP_609877.2 | 2 - 3 | 2007 | 127 | |
GenBank mRNA | BT100034.1 | - | 2 - 3 | 1977 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |