Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273610.1 | NP_001260539.1 | 5 - 6 | 1481 | 94 | |
NM_001273611.1 | NP_001260540.1 | 4 - 5 | 757 | 94 | ||
NM_136037.4 | NP_609881.3 | 5 - 6 | 1503 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |