Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273610.1 | NP_001260539.1 | 8 - 9 | 4502 | 91 | |
NM_001273611.1 | NP_001260540.1 | 7 - 8 | 3778 | 91 | ||
NM_136037.4 | NP_609881.3 | 8 - 9 | 4524 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |