Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo28B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144373.4 | NP_652630.2 | 5 - 6 | 3127 | 95 | |
NM_164758.2 | NP_723294.1 | 5 - 6 | 2982 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |