Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103834.4 | NP_001097304.2 | 2 - 3 | 1015 | 70 | |
NM_001103835.2 | NP_001097305.1 | 2 - 3 | 1015 | 70 | ||
NM_001103836.4 | NP_001097306.2 | 2 - 3 | 1015 | 70 | ||
NM_001144194.2 | NP_001137666.1 | 2 - 3 | 1015 | 70 | ||
NM_001299479.1 | NP_001286408.1 | 2 - 3 | 1015 | 70 | ||
NM_001299480.1 | NP_001286409.1 | 2 - 3 | 1015 | 70 | ||
NM_001316419.1 | NP_001303348.1 | 2 - 3 | 1015 | 70 | ||
NM_079014.3 | NP_523738.2 | 2 - 3 | 1015 | 70 | ||
NM_166034.2 | NP_725362.1 | 2 - 3 | 1015 | 70 | ||
NM_166035.2 | NP_725363.1 | 3 - 4 | 838 | 70 | ||
NM_166036.2 | NP_725364.1 | 3 - 4 | 823 | 70 | ||
GenBank mRNA | AF276664.1 | - | 2 - 3 | 1034 | 70 | |
BT023511.1 | - | 2 - 3 | 1030 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |