Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103834.4 | NP_001097304.2 | 8 - 9 | 2153 | 100 | |
NM_001103835.2 | NP_001097305.1 | 8 - 9 | 2129 | 100 | ||
NM_001103836.4 | NP_001097306.2 | 8 - 9 | 2189 | 100 | ||
NM_001144194.2 | NP_001137666.1 | 8 - 9 | 2153 | 100 | ||
NM_001299479.1 | NP_001286408.1 | 7 - 8 | 1829 | 100 | ||
NM_001299480.1 | NP_001286409.1 | 8 - 9 | 2174 | 100 | ||
NM_001316419.1 | NP_001303348.1 | 7 - 8 | 1874 | 100 | ||
NM_079014.3 | NP_523738.2 | 7 - 8 | 1850 | 100 | ||
NM_166034.2 | NP_725362.1 | 8 - 9 | 2153 | 100 | ||
NM_166035.2 | NP_725363.1 | 8 - 9 | 1673 | 100 | ||
NM_166036.2 | NP_725364.1 | 8 - 9 | 1658 | 100 | ||
GenBank mRNA | AF276664.1 | - | 7 - 8 | 1869 | 100 | |
BT023511.1 | - | 7 - 8 | 1865 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |