Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299482.1 | NP_001286411.1 | 5 - 6 | 789 | 120 | |
NM_001299483.1 | NP_001286412.1 | 4 - 5 | 672 | 120 | ||
NM_001299484.1 | NP_001286413.1 | 4 - 5 | 672 | 120 | ||
NM_137097.3 | NP_610941.1 | 4 - 5 | 672 | 120 | ||
NM_166040.2 | NP_725369.1 | 5 - 6 | 789 | 120 | ||
GenBank mRNA | AY058366.1 | - | 4 - 5 | 646 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |