Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259392.2 | NP_001246321.1 | 4 - 5 | 1597 | 64 | |
NM_137113.4 | NP_610957.1 | 4 - 5 | 1604 | 64 | ||
NM_166048.1 | NP_725382.1 | 3 - 4 | 1724 | 64 | ||
NM_166049.1 | NP_725383.1 | 4 - 5 | 1601 | 64 | ||
GenBank mRNA | AY052097.1 | - | 3 - 4 | 1582 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |