Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014527.2 | NP_001014527.1 | 6 - 7 | 2871 | 61 | |
NM_001274040.1 | NP_001260969.1 | 7 - 8 | 2934 | 61 | ||
NM_001274041.1 | NP_001260970.1 | 6 - 7 | 3335 | 61 | ||
NM_166057.2 | NP_725398.1 | 7 - 8 | 3200 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |