Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299498.1 | NP_001286427.1 | 4 - 5 | 1719 | 91 | |
NM_166063.2 | NP_725407.1 | 4 - 5 | 1719 | 91 | ||
GenBank mRNA | AY084178.1 | - | 4 - 5 | 1718 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |