Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137199.3 | NP_611043.1 | 2 - 3 | 975 | 80 | |
NM_166099.2 | NP_725472.1 | 1 - 2 | 955 | 80 | ||
GenBank mRNA | AF218236.1 | - | 1 - 2 | 739 | 80 | |
AY113411.1 | - | 2 - 3 | 972 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |