Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058010.4 | NP_477358.1 | 4 - 5 | 1186 | 63 | |
NM_166102.2 | NP_725476.1 | 5 - 6 | 1198 | 63 | ||
GenBank mRNA | AF044734.1 | - | 4 - 5 | 952 | 63 | |
AY058794.1 | - | 4 - 5 | 1161 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |