Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103862.2 | NP_001097332.1 | 2 - 3 | 438 | 69 | |
NM_137219.3 | NP_611063.1 | 2 - 3 | 227 | 69 | ||
NM_166118.2 | NP_725495.1 | 2 - 3 | 234 | 69 | ||
NM_166119.2 | NP_725496.1 | 2 - 3 | 293 | 69 | ||
GenBank mRNA | AY051430.1 | - | 2 - 3 | 259 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |