Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103862.2 | NP_001097332.1 | 4 - 5 | 703 | 66 | |
NM_137219.3 | NP_611063.1 | 4 - 5 | 492 | 66 | ||
NM_166118.2 | NP_725495.1 | 4 - 5 | 499 | 66 | ||
NM_166119.2 | NP_725496.1 | 4 - 5 | 558 | 66 | ||
GenBank mRNA | AY051430.1 | - | 4 - 5 | 524 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |