Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137220.3 | NP_611064.2 | 3 - 4 | 2267 | 76 | |
NM_166121.2 | NP_725499.2 | 2 - 3 | 2774 | 76 | ||
NM_176181.2 | NP_788361.1 | 3 - 4 | 2480 | 76 | ||
GenBank mRNA | BT150416.1 | - | 2 - 3 | 2774 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |