Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299540.1 | NP_001286469.1 | 6 - 7 | 3497 | 132 | |
NM_137220.3 | NP_611064.2 | 6 - 7 | 3500 | 132 | ||
NM_166121.2 | NP_725499.2 | 5 - 6 | 4007 | 132 | ||
NM_176181.2 | NP_788361.1 | 6 - 7 | 3713 | 132 | ||
GenBank mRNA | BT150416.1 | - | 5 - 6 | 4007 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |