Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202020.1 | NP_001188949.1 | 4 - 5 | 1587 | 67 | |
NM_001202026.2 | NP_001188955.2 | 18 - 19 | 21802 | 67 | ||
NM_001274089.1 | NP_001261018.1 | 18 - 19 | 21802 | 67 | ||
NM_001274090.1 | NP_001261019.1 | 18 - 19 | 22170 | 67 | ||
NM_166129.3 | NP_725510.2 | 18 - 19 | 22170 | 67 | ||
GenBank mRNA | AY113646.1 | - | 4 - 5 | 1588 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |