Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079032.4 | NP_523756.1 | 11 - 12 | 3156 | 112 | |
NM_166138.5 | NP_725523.3 | 10 - 11 | 2907 | 112 | ||
NM_176183.3 | NP_788363.1 | 12 - 13 | 3951 | 112 | ||
NM_176184.4 | NP_788364.2 | 11 - 12 | 3702 | 112 | ||
GenBank mRNA | AF461100.1 | - | 10 - 11 | 3054 | 112 | |
AF461101.1 | - | 10 - 11 | 2508 | 112 | ||
AY069076.1 | - | 11 - 12 | 3129 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |