Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080084.3 | NP_524823.1 | 8 - 9 | 2361 | 69 | |
NM_166146.2 | NP_725532.1 | 8 - 9 | 2361 | 69 | ||
NM_166147.2 | NP_725533.1 | 7 - 8 | 2099 | 69 | ||
GenBank mRNA | AF212366.1 | - | 8 - 9 | 2325 | 69 | |
AF212367.1 | - | 8 - 9 | 2325 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |