Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168929.2 | NP_730669.1 | 2 - 3 | 404 | 64 | |
GenBank mRNA | BT023364.1 | - | 2 - 3 | 382 | 64 | |
BT029397.1 | - | 2 - 3 | 324 | 64 | ||
KX531694.1 | - | 2 - 3 | 324 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |