Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259947.1 | NP_001246876.1 | 10 - 11 | 2177 | 83 | |
NM_001275268.1 | NP_001262197.1 | 10 - 11 | 1955 | 83 | ||
NM_001275269.1 | NP_001262198.1 | 10 - 11 | 1929 | 83 | ||
NM_001316446.1 | NP_001303375.1 | 10 - 11 | 2011 | 83 | ||
NM_079484.6 | NP_524208.1 | 8 - 9 | 1924 | 83 | ||
NM_168946.4 | NP_730700.2 | 10 - 11 | 1955 | 83 | ||
NM_206423.5 | NP_996145.1 | 9 - 10 | 1858 | 83 | ||
GenBank mRNA | AY060403.1 | - | 9 - 10 | 1806 | 83 | |
X99340.1 | - | 2679 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |