Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPoCk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104196.3 | NP_001097666.1 | 2 - 3 | 1982 | 70 | |
NM_001170025.1 | NP_001163496.1 | 3 - 4 | 2485 | 70 | ||
NM_001259958.2 | NP_001246887.1 | 3 - 4 | 2299 | 70 | ||
NM_001370004.1 | NP_001356985.1 | 2 - 3 | 1899 | 70 | ||
NM_168961.5 | NP_730742.1 | 2 - 3 | 2267 | 70 | ||
NM_168962.4 | NP_730744.1 | 3 - 4 | 1956 | 70 | ||
NM_168963.4 | NP_730745.1 | 2 - 3 | 1899 | 70 | ||
GenBank mRNA | BT082029.1 | - | 3 - 4 | 2485 | 70 | |
BT133078.1 | - | 3 - 4 | 1946 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |