Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275304.1 | NP_001262233.1 | 3 - 4 | 366 | 73 | |
NM_143697.5 | NP_651954.3 | 3 - 4 | 366 | 73 | ||
NM_168984.5 | NP_730773.4 | 3 - 4 | 377 | 73 | ||
GenBank mRNA | AY070910.1 | - | 3 - 4 | 364 | 73 | |
U49721.1 | - | 3 - 4 | 356 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |